Skip to Content
Merck
All Photos(2)

Key Documents

910910

Sigma-Aldrich

5-Deoxy-D-ribose

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$554.00

$554.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$554.00

About This Item

Empirical Formula (Hill Notation):
C5H10O4
CAS Number:
Molecular Weight:
134.13
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

$554.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

viscous liquid

storage temp.

−20°C

SMILES string

O[C@H]([C@H](O)C)[C@@H](O)C=O

InChI

1S/C5H10O4/c1-3(7)5(9)4(8)2-6/h2-5,7-9H,1H3/t3-,4+,5-/m1/s1

InChI key

WDRISBUVHBMJEF-MROZADKFSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service