855448
D-Leucine
99%, for peptide synthesis, ReagentPlus®
Synonym(s):
(R)-2-Amino-4-methylpentanoic acid
About This Item
Recommended Products
product name
D-Leucine, ReagentPlus®, 99%
Quality Level
product line
ReagentPlus®
Assay
99%
form
powder (crystals)
optical activity
[α]23/D −14.7°, c = 2 in 5 M HCl
optical purity
ee: 97% (GLC)
reaction suitability
reaction type: solution phase peptide synthesis
mp
>300 °C (lit.)
application(s)
peptide synthesis
SMILES string
CC(C)C[C@@H](N)C(O)=O
InChI
1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m1/s1
InChI key
ROHFNLRQFUQHCH-RXMQYKEDSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Chromatograms
application for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service