Skip to Content
Merck
All Photos(1)

Key Documents

90526

Supelco

Capsiate

analytical standard

Synonym(s):

(6E)-8-Methyl-6-nonenoic acid (4-hydroxy-3-methoxyphenyl)methyl ester, 4-Hydroxy-3-methoxybenzyl (E)-8-methyl-6-nonenoate, Vanillyl (E)-8-methyl-6-nonenoate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$638.00

$638.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$638.00

About This Item

Empirical Formula (Hill Notation):
C18H26O4
CAS Number:
Molecular Weight:
306.40
Beilstein:
7933697
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

$638.00


Estimated to ship onApril 17, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O(C)c1c(ccc(c1)COC(=O)CCCC\C=C\C(C)C)O

InChI

1S/C18H26O4/c1-14(2)8-6-4-5-7-9-18(20)22-13-15-10-11-16(19)17(12-15)21-3/h6,8,10-12,14,19H,4-5,7,9,13H2,1-3H3/b8-6+

InChI key

ZICNYIDDNJYKCP-SOFGYWHQSA-N

General description

Capsiate is a capsaicinoid and a non-pungent ingredient of CH-19 sweet pepper. It exhibits anti-obesity and anti-inflammatory properties. Capsiate is reported to be an agonist of capsaicin?binding transient potential vanilloid 1 (TRPV 1) receptors.[1][2][3]

Application

Capsiate may be used as an analytical standard for the determination of the analyte in Capsicum species[4][5] by chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Capsiate, a non-pungent capsaicin analog, reduces body fat without weight rebound like swimming exercise in mice
Haramizu S, et al.
Biomedical Research, 32(4), 279-284 (2011)
Determination of capsinoids by HPLC-DAD in Capsicum species
Singh S, et al.
Journal of Agricultural and Food Chemistry, 57(9), 3452-3457 (2009)
Quantitation of capsiate and dihydrocapsiate and tentative identification of minor capsinoids in pepper fruits (Capsicum spp.) by HPLC-ESI-MS/MS (QTOF)
Fayos O, et al.
Food Chemistry, 270(9), 264-272 (2019)
Different TRPV 1-mediated brain responses to intragastric infusion of capsaicin and capsiate
Tsurugizawa T, et al.
European Journal of Neurology, 38(11), 3628-3635 (2013)
Capsiate inhibits ultraviolet B-induced skin inflammation by inhibiting Src family kinases and epidermal growth factor receptor signaling
Lee EJ, et al.
Free Radical Biology & Medicine, 48(9), 1133-1143 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service