Skip to Content
Merck
All Photos(3)

Key Documents

M1022

Sigma-Aldrich

Dihydrocapsaicin

from Capsicum sp., ≥85% (HPLC), powder, VR1 vanilloid receptor agonist

Synonym(s):

6,7-Dihydrocapsaicin, 8-Methyl-N-vanillylnonanamide, N-[(4-Hydroxy-3-methoxyphenyl)methyl]-8-methylnonenamide

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$191.00
250 MG
$554.00
1 G
$1,670.00

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$191.00
250 MG
$554.00
1 G
$1,670.00

About This Item

Empirical Formula (Hill Notation):
C18H29NO3
CAS Number:
Molecular Weight:
307.43
Beilstein:
2815150
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$191.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Dihydrocapsaicin, from Capsicum sp., ≥85%

biological source

Capsicum sp.

Assay

≥85%

storage temp.

2-8°C

SMILES string

COc1cc(CNC(=O)CCCCCCC(C)C)ccc1O

InChI

1S/C18H29NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h10-12,14,20H,4-9,13H2,1-3H3,(H,19,21)

InChI key

XJQPQKLURWNAAH-UHFFFAOYSA-N

Gene Information

human ... TRPV1(7442)
rat ... Trpv4(66026)

Looking for similar products? Visit Product Comparison Guide

Application

Dihydrocapsaicin has been used:
  • as a standard to analyze the level of dihydrocapsaicin in endogenous An2 and capsaicin synthase silenced fruits[1]
  • as an external standard to determine the variability in capsaicinoid content in different landraces of capsicum[2]
  • in the activation or ablation of capsaicin-sensitive primary afferents (CSPA) fibres[3]

Biochem/physiol Actions

Dihydrocapsaicin/8-methyl-N-vanillylnonanamide; N-[−4-hydroxy-3-methoxybenzyl] 8-methylnonanamide (DHC) is an active component of capsaicinoids in chili peppers.[4] DHC is capable of reducing body temperature. Hence it is considered as a pharmaceutical-induced hypothermia (PIH) candidate to treat cardiac arrest (CA) patients.[5] It may possess anti-cancer, anti-inflammation, antioxidant, anti-obesity properties. DHC might help in the reduction of oxidative stress and inflammation in ischemia and reperfusion (I/R) injury. DHC can repress nuclear factor kappa-light-chain-enhancer of activated B cells (NF-κB) signaling pathway.[4]
VR1 vanilloid receptor agonist.

Linkage

Capsaicin analog.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Variability in capsaicinoid content in different landraces of Capsicum cultivated in north-eastern India
Islam Md A, et al.
Sci. Hortic., 183, 66-71 (2015)
Dihydrocapsaicin-induced Hypothermia after Asphyxial Cardiac Arrest in Rats
He J, et al.
Conference proceedings : Annual International Conference of the IEEE Engineering in Medicine and Biology Society, 2016, 1858-1858 (2016)
Y J Surh et al.
Life sciences, 56(16), PL305-PL311 (1995-03-10)
A new metabolic oxidation pathway of capsaicin (N-[(4-hydroxy-3-methoxyphenyl)-methyl]-8-methyl-(E)-6 -nonenamide), a major pungent and pharmacologically active principle of hot peppers, was investigated. Incubation of capsaicin with phenobarbital-induced rat liver postmitochondrial supernatant enriched with NADPH-generating system produced N-(4,5-dihydroxy-3-methoxybenzyl)-(E)-6 -nonenylamide and a more
Dihydrocapsaicin Attenuates Blood Brain Barrier and Cerebral Damage in Focal Cerebral Ischemia/Reperfusion via Oxidative Stress and Inflammatory
Janyou A, et al.
Scientific Reports, 7 (2017)
Harnessing anthocyanin-rich fruit: a visible reporter for tracing virus-induced gene silencing in pepper fruit
Kim J, et al.
Plant methods, 13(1), 1-10 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service