Skip to Content
Merck
All Photos(1)

Key Documents

89787

Sigma-Aldrich

(Trimethylsiloxy)ethylene

Wacker Chemie AG, ≥96% (GC)

Synonym(s):

TMSO-ethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOCH=CH2
CAS Number:
Molecular Weight:
116.23
Beilstein:
1738846
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥96% (GC)

form

liquid

manufacturer/tradename

Wacker Chemie AG

refractive index

n20/D 1.389 (lit.)

bp

74 °C (lit.)

density

0.779 g/mL at 25 °C (lit.)

SMILES string

[H]\C([H])=C(\[H])O[Si](C)(C)C

InChI

1S/C5H12OSi/c1-5-6-7(2,3)4/h5H,1H2,2-4H3

InChI key

HFTNNOZFRQLFQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

prices for bulk quantities on request

replaced by

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service