Skip to Content
Merck
All Photos(1)

Key Documents

235040

Sigma-Aldrich

1-Phenyl-1-trimethylsiloxyethylene

98%

Synonym(s):

α-(Trimethylsiloxy)styrene, Acetophenone enol trimethylsilyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$153.00
25 G
$711.00

$153.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$153.00
25 G
$711.00

About This Item

Linear Formula:
C6H5C(=CH2)OSi(CH3)3
CAS Number:
Molecular Weight:
192.33
Beilstein:
1306914
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$153.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

>1 (vs air)

Assay

98%

form

liquid

refractive index

n20/D 1.502 (lit.)

bp

88-89 °C/11 mmHg (lit.)

density

0.938 g/mL at 25 °C (lit.)

functional group

phenyl

storage temp.

2-8°C

SMILES string

C[Si](C)(C)OC(=C)c1ccccc1

InChI

1S/C11H16OSi/c1-10(12-13(2,3)4)11-8-6-5-7-9-11/h5-9H,1H2,2-4H3

InChI key

AFFPCIMDERUIST-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Phenyl-1-trimethylsiloxyethylene is a styrene type silyl enol ether, reacts with formaldehyde and 2,4-pentanedione to yield the corresponding dihydropyran[1]. It also undergoes Mukaiyama aldol reaction with aldehydes in water in the presence of amphiphilic calix[6]arene derivatives as surfactants[2].

Application

1-Phenyl-1-trimethylsiloxyethylene has been used in the synthesis of:
  • β-amino ketones in water via Mannich-type reaction[3]
  • cis-2,6-disubstituted dihydropyrans via three-component, one-pot cascade reaction[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Calix [6] arene derivatives bearing sulfonate and alkyl groups as surfactants in Sc (OTf)3-catalyzed Mukaiyama aldol reactions in water.
Tian H-Y, et al.
Tetrahedron Letters, 41(15), 2529-2532 (2000)
Novel one-pot Mannich-type reaction in water: indium trichloride-catalyzed condensation of aldehydes, amines and silyl enol ethers for the synthesis of ?-amino ketones and esters.
Loh T-P and Wei L-L.
Tetrahedron Letters, 39(3), 323-326 (1998)
Catalyst-free aqueous multicomponent domino reactions from formaldehyde and 1, 3-dicarbonyl derivatives.
Gu Y, et al.
Green Chemistry, 11(12), 1968-1972 (2009)
Robert J Hinkle et al.
Tetrahedron, 65(34), 6834-6839 (2010-02-18)
The rapid synthesis of cis-2,6-disubstituted dihydropyrans is achieved in a three-component, one-pot cascade reaction. BiBr(3)-ediated addition of ketene silyl acetals or silyl enol ethers to beta,gamma-unsaturated cis-4-trimethylsilyl-3-butenal provides a Mukaiyama aldol adduct containing a vinylsilane moiety tethered to a silyl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service