Skip to Content
Merck
All Photos(1)

Key Documents

87976

Supelco

Polyethylene glycol Standard ReadyCal Set Mp ~250-45′000

for GPC

Synonym(s):

PEG Molecular Weight Calibration Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$795.00

$795.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Select a Size

Change View
1 SET
$795.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$795.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

grade

for GPC

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

gel permeation chromatography (GPC): suitable

format

neat

storage temp.

2-8°C

Components

ReadyCal Standards are polymer cocktails pre-weighed into autosampler vials for the convenient calibration of GPC/SEC/GFC systems.
This kit contains 10 sets of 3 color-coded vials, each containing four polyethylene glycols of different molar masses. Molar masses range from ~250 to ~45,000 Da and are carefully selected so that polymer traces are baseline separated.
This kit contains sufficient material for at least 10 calibrations and includes an instruction manual and calibration curve with measurement conditions.
Suitable for calibrations in aqueous solutions, DMF and DMAc (THF only for low molar masses).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service