Skip to Content
Merck
All Photos(1)

Key Documents

87708

Supelco

Tetramethylammonium bromide

suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(Br)
CAS Number:
Molecular Weight:
154.05
Beilstein:
3620955
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21
form:
crystals
Assay:
≥99.0% (AT)

description

cationic

Quality Level

Assay

≥99.0% (AT)

form

crystals

quality

LiChropur

technique(s)

ion pair chromatography: suitable

mp

>300 °C (lit.)

λ

10 % in H2O

UV absorption

λ: 240 nm Amax: 0.06
λ: 250 nm Amax: 0.03
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for filter test

SMILES string

[Br-].C[N+](C)(C)C

InChI

1S/C4H12N.BrH/c1-5(2,3)4;/h1-4H3;1H/q+1;/p-1

InChI key

DDFYFBUWEBINLX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Ion-pairing reagents, when used as mobile phase additives, promote the analysis of ionic and highly polar substances on reversed phase HPLC columns. The purity of mobile phase additives is of highest significance for their effective application. Sigma-Aldrich makes available a variety of tailor-made reagents for anionic (quaternary ammonium and phosphonium salts) and cationic (alkanesulfonates) separations. All mobile phase additives are tested thoroughly with special consideration on the demands of modern reversed phase HPLC:

Application


  • pH Standardization in Analytical Chemistry: Buffers serve as critical components in the batch-to-batch reproducibility studies for primary pH methods, ensuring high precision in pH measurement across various scientific disciplines, from pharmaceuticals to environmental science (Asakai et al., 2024).



Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Chong et al.
Langmuir : the ACS journal of surfaces and colloids, 28(4), 2091-2098 (2011-11-08)
A new water-soluble conjugated polymer containing fluorene and boron-dipyrromethene repeat units in the backbones (PBF) that exhibits red emission was synthesized and characterized. Cationic PBF forms uniform nanoparticles with negatively charged disodium salt 3,3'-dithiodipropionic acid (SDPA) in aqueous solution through
Qin Chen et al.
Bioresource technology, 117, 186-192 (2012-05-23)
The aim of this study was to explore the synergies of laccase, a ligninolytic enzyme, with cellulose and hemicellulase amendments on ensiled corn stover. Molecular signals of lignin decomposition were observed by tetramethylammonium hydroxide thermochemolysis and gas chromatography-mass spectroscopy (TMAH-GC-MS)
Maggie Tam et al.
The Analyst, 136(15), 3098-3106 (2011-06-24)
A novel analytical method, called Liquid Phase Ion Mobility Spectrometry (LiPIMS) was demonstrated, where aqueous phase analytes were ionized and introduced into non-aqueous liquids, transported by an external electric field from the point of generation to a collection electrode. Ions
Lina Barreto et al.
Eukaryotic cell, 10(9), 1241-1250 (2011-07-05)
Potassium homeostasis is crucial for living cells. In the yeast Saccharomyces cerevisiae, the uptake of potassium is driven by the electrochemical gradient generated by the Pma1 H(+)-ATPase, and this process represents a major consumer of the gradient. We considered that
Konrad Rykaczewski et al.
Langmuir : the ACS journal of surfaces and colloids, 29(3), 881-891 (2012-12-25)
The prospect of enhancing the condensation rate by decreasing the maximum drop departure diameter significantly below the capillary length through spontaneous drop motion has generated significant interest in condensation on superhydrophobic surfaces (SHS). The mobile coalescence leading to spontaneous drop

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service