Merck
  • Multimode multidrop serial coalescence effects during condensation on hierarchical superhydrophobic surfaces.
Multimode multidrop serial coalescence effects during condensation on hierarchical superhydrophobic surfaces.
Langmuir : the ACS journal of surfaces and colloids (2012-12-25)
Konrad Rykaczewski, Adam T Paxson, Sushant Anand, Xuemei Chen, Zuankai Wang, Kripa K Varanasi
ABSTRACT
The prospect of enhancing the condensation rate by decreasing the maximum drop departure diameter significantly below the capillary length through spontaneous drop motion has generated significant interest in condensation on superhydrophobic surfaces (SHS). The mobile coalescence leading to spontaneous drop motion was initially reported to occur only on hierarchical SHS, consisting of both nanoscale and microscale topological features. However, subsequent studies have shown that mobile coalescence also occurs on solely nanostructured SHS. Thus, recent focus has been on understanding the condensation process on nanostructured surfaces rather than on hierarchical SHS. In this work, we investigate the impact of microscale topography of hierarchical SHS on the droplet coalescence dynamics and wetting states during the condensation process. We show that isolated mobile and immobile coalescence between two drops, almost exclusively focused on in previous studies, are rare. We identify several new droplet shedding modes, which are aided by tangential propulsion of mobile drops. These droplet shedding modes comprise of multiple droplets merging during serial coalescence events, which culminate in formation of a drop that either departs or remains anchored to the surface. We directly relate postmerging drop adhesion to formation of drops in nanoscale as well as microscale Wenzel and Cassie-Baxter wetting states. We identify the optimal microscale feature spacing of the hierarchical SHS, which promotes departure of the highest number of microdroplets. This optimal surface architecture consists of microscale features spaced close enough to enable transition of larger droplets into micro-Cassie state yet, at the same time, provides sufficient spacing in-between the features for occurrence of mobile coalescence.
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Supelco
Tetramethylammonium sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Tetramethylammonium chloride, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Supelco
Tetramethylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium chloride, reagent grade, ≥98%
Sigma-Aldrich
Tetramethylammonium fluoride, 97%
Sigma-Aldrich
Tetramethylammonium chloride solution, Molecular Biology
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥97%
Sigma-Aldrich
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
Supelco
Tetramethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Tetramethylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Tetramethylammonium bromide, 98%
Sigma-Aldrich
Tetramethylammonium iodide, 99%