Skip to Content
Merck
All Photos(1)

Key Documents

86618

Supelco

Tetraethylammonium tetrafluoroborate

for electrochemical analysis, ≥99.0%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$126.00
25 G
$497.00

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$126.00
25 G
$497.00

About This Item

Linear Formula:
(C2H5)4N(BF4)
CAS Number:
Molecular Weight:
217.06
Beilstein:
3917638
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61
form:
crystals
solubility:
acetonitrile: 0.1 g/mL, clear, colorless

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.0% (T)
≥99.0%

form

crystals

mp

≥300 °C (lit.)

solubility

acetonitrile: 0.1 g/mL, clear, colorless

SMILES string

F[B-](F)(F)F.CC[N+](CC)(CC)CC

InChI

1S/C8H20N.BF4/c1-5-9(6-2,7-3)8-4;2-1(3,4)5/h5-8H2,1-4H3;/q+1;-1

InChI key

XJRAKUDXACGCHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application


  • High-rate electrochemical capacitors based on ordered mesoporous silicon carbide-derived carbon: This study discusses the use of Tetraethylammonium tetrafluoroborate in enhancing the performance of high-rate electrochemical capacitors, focusing on ordered mesoporous silicon carbide-derived carbon structures (Korenblit et al., 2010).

  • Exactly matched pore size for the intercalation of electrolyte ions determined using the tunable swelling of graphite oxide in supercapacitor electrodes: The application of Tetraethylammonium tetrafluoroborate in electrolyte solutions for supercapacitors, demonstrating how it facilitates the exact matching of pore sizes in graphite oxide electrodes for optimal ion intercalation (Sun et al., 2018).

  • A universal model for nanoporous carbon supercapacitors applicable to diverse pore regimes, carbon materials, and electrolytes: Describes a modeling approach that incorporates the characteristics of Tetraethylammonium tetrafluoroborate in various carbon materials and electrolytes, enhancing the understanding and design of supercapacitors (Huang et al., 2008).

  • Sub-micrometer Novolac-Derived Carbon Beads for High Performance Supercapacitors and Redox Electrolyte Energy Storage: Research on utilizing Tetraethylammonium tetrafluoroborate in novel sub-micrometer novolac-derived carbon beads, which significantly boost the efficiency and energy storage capacity of supercapacitors (Krüner et al., 2016).

  • Biomass-based Hierarchical Porous Carbon for Supercapacitors: Effect of Aqueous and Organic Electrolytes on the Electrochemical Performance: Explores the efficacy of Tetraethylammonium tetrafluoroborate in enhancing the electrochemical performance of biomass-derived hierarchical porous carbons in various electrolyte environments (Chen et al., 2019).

Other Notes

Supporting electrolyte[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Fruianu et al.
J. Chem. Soc. Perkin Trans. II, 2039-2039 (1994)
Y. Matsumura et al.
Tetrahedron, 51, 6411-6411 (1995)
Kecheng Li et al.
Scientific reports, 6, 39689-39689 (2016-12-22)
Graphene film has been demonstrated as promising active materials for electric double layer capacitors (EDLCs), mainly due to its excellent mechanical flexibility and freestanding morphology. In this work, the distribution and variation pattern of electrolyte ions in graphene-film based EDLC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service