Skip to Content
Merck
All Photos(3)

Key Documents

765945

Sigma-Aldrich

Tetramethylammonium bis(trifluoromethanesulfonyl)imide

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$145.00
5 G
$581.00

$145.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$145.00
5 G
$581.00

About This Item

Empirical Formula (Hill Notation):
C6H12F6N2O4S2
CAS Number:
Molecular Weight:
354.29
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

$145.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

mp

132-136 °C

storage temp.

20-25°C

SMILES string

C[N+](C)(C)C.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C4H12N.C2F6NO4S2/c1-5(2,3)4;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h1-4H3;/q+1;-1

InChI key

JTCSZQBQVDPVMT-UHFFFAOYSA-N

Related Categories

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service