Skip to Content
Merck
All Photos(1)

Key Documents

81395

Supelco

Poly(vinyl chloride) carboxylated

Selectophore, 1.8% carboxyl basis

Synonym(s):

Poly(vinyl chloride-co-acrylic acid)

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
$358.00

$358.00


Estimated to ship onMay 09, 2025


Request a Bulk Order

Select a Size

Change View
50 G
$358.00

About This Item

Linear Formula:
(C3H4O2 · C2H3Cl)x
CAS Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

$358.00


Estimated to ship onMay 09, 2025


Request a Bulk Order

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

form

powder

concentration

1.8% (carboxyl)

SMILES string

ClC=C

InChI

1S/C3H4O2.C2H3Cl/c1-2-3(4)5;1-2-3/h2H,1H2,(H,4,5);2H,1H2

InChI key

SQNNHEYXAJPPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Application

Poly(vinyl chloride) carboxylated may be used in the preparation of ultrafiltration membranes using phase-inversion method.[1]

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation, structure, and transport properties of ultrafiltration membranes of poly (vinyl chloride)(PVC), carboxylated poly (vinyl chloride)(CPVC), and PVC/CPVC blends.
Babu R P and Gaikar G V.
Journal of Applied Polymer Science, 73(7), 1117-1130 (1999)
IIIrd International Symposium on Neural Transplantation. Cambridge, 6-11 August, 1989. Proceedings.
Progress in brain research, 82, 1-734 (1990-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service