Skip to Content
Merck
Poly(vinyl chloride) carboxylated Selectophore™, 1.8% carboxyl basis

Poly(vinyl chloride) carboxylated

Poly(vinyl chloride) carboxylated Selectophore™, 1.8% carboxyl basis
Synonym(s):
Poly(vinyl chloride-co-acrylic acid)
Linear Formula:
(C3H4O2 · C2H3Cl)x
CAS Number:

Products

Brand
Physical Form
Product Line
Quality Segment
Polymer Composition
Polymer Type