Skip to Content
Merck
All Photos(1)

Key Documents

80957

Millipore

Plate Count Skim Milk Agar

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

form

powder

Quality Level

shelf life

limited shelf life, expiry date on the label

composition

agar, 10.5 g/L
casein peptone, 5 g/L
D(+)-glucose, 1 g/L
skim milk powder (inhibitor-free), 1 g/L
yeast extract, 2.5 g/L

final pH

7.0±0.2 (25 °C)

application(s)

environmental
food and beverages

microbiology

suitability

nonselective for Bacillus spp.
nonselective for Candida spp.
nonselective for Escherichia coli
nonselective for Lactobacillus spp.
nonselective for Lactococcus spp.
nonselective for Listeria spp.
nonselective for Pseudomonas spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms

Application

For the enumeration of bacteria in milk and dairy products.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Terplan et al.
Archiv Fur Lebensmittelhygiene, 18, 9-9 (1967)

Articles

Cost-effective molecular biology method for lactobacilli detection is presented as an alternative to PCR-based methods.

An article concerning the detection, identification, differentiation, and cultivation of Pseudomonas species.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service