Skip to Content
Merck
All Photos(1)

Documents

1.15363

Millipore

Skim milk powder

suitable for microbiology

Synonym(s):

Skim milk powder, Dry skim milk, non-fat milk solids, skimmed milk powder,

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41106212
EC Index Number:
310-127-6
NACRES:
NA.74

biological source

bovine milk

Quality Level

form

solid

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 500 g

pH

6-8 (20 °C, 20 g/L in H2O)

solubility

700 g/L

bulk density

500 kg/m3

application(s)

microbiology
pathogen testing

storage temp.

15-25°C

General description

Skim milk powder is widely used in microbiology as a nutrient source in microbial culture media. It provides a complex mixture of nutrients, including protein, lactose, and minerals, that support the growth of a variety of microorganisms. Skim milk powder is commonly used in media formulations for the cultivation of lactic acid bacteria, including Streptococcus and Lactobacillus species. It can also be used as a supplement in media for the growth of other microbial cells, including fungi, yeasts, and some bacteria that utilize milk proteins as a nutrient source. Additionally, skim milk powder has the advantage of being shelf-stable and easily rehydratable, making it a convenient and cost-effective source of nutrients for microbial cultivation.

Application

Skim milk powder is usually used as an ingredient in many microbial culture media for the growth and identification of bacteria, yeasts, and molds from various pharmaceutical, environmental, food and beverage samples. Furthermore, it is described in the EN ISO 6887-4:2017 to be used as pre-enrichment broth for the detection of Salmonella spp. and STEC when testing cocoa and cocoa containing products (use 100 g/l; sterilize after reconstitution). As an ingredient in culture media Skim milk powder is usually employed in concentrations of 0.1 to 1.0%.

Linkage

Replaces: 1.15363.9017

Analysis Note

Assay protein (ex N, calc. on dried substance): ≥ 35 %
Lactose (enzymatically): 40 - 52 %
pH-value (1 %; water): 6 - 7
Fats: ≤ 1.5 %
Free acid (as lactic acid): ≤ 2 %
Ash (800 °C): ≤ 10 %
Water (according to Karl Fischer): ≤ 5.0 %
Suitability for microbiology: passes test
Microbiological test: passes test
Identity (NIR): passes test

Other Notes

Rehydrating the Skim milk powder, or using it for the preparation of culture media, the milk powder should be gently added to water with proper stirring to make sure that no clumps are present in the rehydrated skim milk or the prepared media

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service