Skip to Content
Merck
All Photos(1)

Key Documents

80725

Supelco

Hydrocinnamic acid

analytical standard

Synonym(s):

3-Phenylpropionic acid, Benzylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2COOH
CAS Number:
Molecular Weight:
150.17
Beilstein:
907515
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

bp

280 °C (lit.)

mp

45-48 °C (lit.)

density

1.071 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

OC(=O)CCc1ccccc1

InChI

1S/C9H10O2/c10-9(11)7-6-8-4-2-1-3-5-8/h1-5H,6-7H2,(H,10,11)

InChI key

XMIIGOLPHOKFCH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katheryn M Goodrich et al.
Journal of agricultural and food chemistry, 62(46), 11190-11199 (2014-10-23)
Metabolism of flavanols (catechins, procyanidins) by gut microbiota has been extensively characterized. Comparatively little is known about accumulation of flavanols and their metabolites in the colon tissues, particularly during chronic exposure to low doses. Mice were fed low doses of
Shiraz Tyebji et al.
Neurobiology of disease, 74, 41-57 (2014-12-03)
Stimulation of dopamine D1 receptor (D1R) and adenosine A2A receptor (A2AR) increases cAMP-dependent protein kinase (PKA) activity in the brain. In Huntington's disease, by essentially unknown mechanisms, PKA activity is increased in the hippocampus of mouse models and patients and
Jong Min Lee et al.
Biomaterials, 39, 173-181 (2014-12-04)
A new design concept in controlled release chemistry is reported in this study. Unlike current depots that release drugs in all direction by an isotropic way, we demonstrate that directional release only to a clinically beneficial direction results in improved

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service