Merck
  • Dietary supplementation with cocoa flavanols does not alter colon tissue profiles of native flavanols and their microbial metabolites established during habitual dietary exposure in C57BL/6J mice.
Dietary supplementation with cocoa flavanols does not alter colon tissue profiles of native flavanols and their microbial metabolites established during habitual dietary exposure in C57BL/6J mice.
Journal of agricultural and food chemistry (2014-10-23)
Katheryn M Goodrich, Melanie R Dorenkott, Liyun Ye, Sean F O'Keefe, Matthew W Hulver, Andrew P Neilson
ABSTRACT
Metabolism of flavanols (catechins, procyanidins) by gut microbiota has been extensively characterized. Comparatively little is known about accumulation of flavanols and their metabolites in the colon tissues, particularly during chronic exposure to low doses. Mice were fed low doses of cocoa flavanols for 12 weeks. Supplementation of the control diet with flavanols did not increase colonic tissue accumulation of flavanols nor microbial metabolites versus control. The type of cocoa flavanols did not affect colonic tissue accumulation of native flavanols or metabolites. Total phenolic content of the diets indicated that these results are not explained by background levels of undetected phenolics in the control diet. This is the longest known chronic flavanol feeding study to examine colonic tissue accumulation. Vast differences appear to exist between acute high doses and chronic low doses, to which gut microbiota and epithelium adapt. These results indicate that the fate of flavanols in the colon during chronic exposure is not fully understood.
Product Description
Supelco
3,4-Dihydroxybenzoic acid, analytical standard
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Supelco
4-Hydroxybenzoic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, ≥99%
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
trans-p-Coumaric acid, analytical standard
Sigma-Aldrich
Phloretin, ≥99%
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
(+)-Catechin hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
(+)-Catechin hydrate, ≥96.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
3-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
3-Phenylpropionic acid, 99%, FG
Sigma-Aldrich
4-Hydroxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
m-Coumaric acid, analytical standard
Ferulic acid, European Pharmacopoeia (EP) Reference Standard
Protocatechuic acid, primary reference standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Vanillic acid, ≥97%, FG