Skip to Content
Merck
All Photos(2)

Key Documents

79879

Supelco

2,2,3,3,3-Pentafluoro-1-propanol

for GC derivatization, LiChropur, ≥98.5% (GC)

Synonym(s):

PFPOH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF2CH2OH
CAS Number:
Molecular Weight:
150.05
Beilstein:
1743133
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.05
Pricing and availability is not currently available.

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

Assay

≥98.5% (GC)

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.288 (lit.)

bp

80 °C/748 mmHg (lit.)

density

1.505 g/mL at 25 °C (lit.)

SMILES string

OCC(F)(F)C(F)(F)F

InChI

1S/C3H3F5O/c4-2(5,1-9)3(6,7)8/h9H,1H2

InChI key

PSQZJKGXDGNDFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Adsorption Mechanisms: Studies the adsorption mechanisms of Pentafluoropropanol (5FP) onto activated carbons produced by the Tetraethoxysilane Sol-Gel method, relevant for environmental and filtration applications (Kawasaki et al., 1997).

  • Cocaine and Metabolites Detection: Details the use of 2,2,3,3,3-Pentafluoro-1-propanol in the analysis of cocaine, its metabolites, and pyrolytic products, highlighting its role in forensic and toxicological studies (Cardona et al., 2006).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT RE 2 Inhalation

Target Organs

Liver

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Callie A Cole et al.
Physical chemistry chemical physics : PCCP, 16(33), 17835-17844 (2014-07-19)
Uracil is an essential biomolecule for terrestrial life, yet its prebiotic formation mechanisms have proven elusive for decades. Meteorites have been shown to contain uracil and the interstellar abundance of aromatic species and nitrogen-containing molecules is well established, providing support

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service