Skip to Content
Merck
All Photos(1)

Key Documents

77264

Sigma-Aldrich

Octadecafluorodecahydronaphthalene

(cis+trans), ≥94% (GC)

Synonym(s):

Perfluorodecahydronaphthalene, Perfluorodecalin

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$272.00
250 ML
$1,130.00

$272.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
50 ML
$272.00
250 ML
$1,130.00

About This Item

Empirical Formula (Hill Notation):
C10F18
CAS Number:
Molecular Weight:
462.08
Beilstein:
2067113
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$272.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Quality Level

Assay

≥94% (GC)

form

liquid

refractive index

n20/D 1.314 (lit.)

bp

141-143 °C (lit.)

density

1.941 g/mL at 20 °C (lit.)
1.941 g/mL at 20 °C

functional group

fluoro

SMILES string

FC1(F)C(F)(F)C(F)(F)C2(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C2(F)C1(F)F

InChI

1S/C10F18/c11-1-2(12,5(17,18)9(25,26)7(21,22)3(1,13)14)6(19,20)10(27,28)8(23,24)4(1,15)16

InChI key

UWEYRJFJVCLAGH-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F

Flash Point(C)

59 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisa Maillard et al.
Biomaterials, 32(35), 9282-9289 (2011-09-09)
Disruption of microenvironment and decrease in oxygen supply during isolation and culture lead to pancreatic islet injury and their poor survival after transplantation. This study aimed to create a matrix for culturing islets, using fibrin as scaffold and perfluorodecalin as
Kavitha K Reddy et al.
Lasers in surgery and medicine, 45(2), 76-80 (2012-12-21)
Laser tattoo removal using multiple passes per session, with each pass delivered after spontaneous resolution of whitening, improves tattoo fading in a 60-minute treatment time. Our objective was to evaluate the safety and efficacy of topical perfluorodecalin (PFD) in facilitating
Steven Garnier et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 251(9), 2081-2085 (2013-03-19)
To assess the anatomic and functional results of 360° retinotomy for rhegmatogenous retinal detachment (RD) with severe proliferative vitreoretinopathy (PVR). Retrospective, non-comparative, interventional, case study of 20 consecutive patients. All surgical operations are as follows: vitrectomy, membrane peeling, PFCL, circumferential
Maciej Pilarek et al.
Microbial cell factories, 10, 50-50 (2011-06-29)
Liquid perfluorochemicals (PFCs) are interesting oxygen carriers in medicine and biotechnology with a high solubility for oxygen. They have been repeatedly used for improving oxygen transfer into prokaryotic and eukaryotic cell cultures, however their application is still limited. Here we
Xabier Murgia et al.
Pediatric research, 72(4), 393-399 (2012-07-17)
Aerosolized perfluorocarbon (PFC) has been proposed as an alternative method of PFC administration; however, the efficacy of aerosolized PFC in a preterm animal model has not yet been demonstrated. Twelve preterm lambs were randomized to two groups: a perfluorodecalin (PFD)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service