Skip to Content
Merck
All Photos(1)

Key Documents

76704

Millipore

modified Tryptic Soy Broth- Dehydrated Culture Media

NutriSelect® Plus, novobiocin 0.02 g/L, according to ISO 16654:2001, powder, selective for enterovirulent E. coli (EEC)

Synonym(s):

Modified Tryptic Soy Broth with Novobiocin, Modified Tryptic Soy Broth; EHEC enrichment broth, mTSB Broth with Novobiocin

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$238.00

$238.00


Estimated to ship onMay 22, 2025


Request a Bulk OrderRequest more information

Select a Size

Change View
500 G
$238.00

About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

$238.00


Estimated to ship onMay 22, 2025


Request a Bulk OrderRequest more information

Agency

according to ISO 16654:2001

Quality Level

sterility

non-sterile

form

powder

shelf life

limited, expiry date on the label

composition

bile salts No. 3, 1.5 g/L
di-potassium hydrogen phosphate, 4.0 g/L
D(+)-glucose, 2.5 g/L
novobiocin, 0.02 g/L
peptone from casein, 17.0 g/L
peptone from soymeal, 3.0 g/L
sodium chloride, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

packaging

pkg of -500 g

storage condition

(Dry place, tightly sealed containers)

technique(s)

microbiological culture: suitable

color

faint brown

final pH

7.3±0.2 (25 °C)

application(s)

agriculture
food and beverages
microbiology
pathogen testing

shipped in

wet ice

storage temp.

2-8°C

suitability

selective for Escherichia coli
selective for coliforms
selective for enterovirulent E. coli (EEC)

Looking for similar products? Visit Product Comparison Guide

General description

mTSB Broth with Novobiocin is suitable for the selective enrichment of enterohemorrhagic Escherichia coli (EHEC) in foods intended for human and animal consumption. The peptone provides amino acids and other complex nitrogenous substances. Glucose is a fermentable carbohydrate. Sodium chloride delivers osmotic balance. Bile salts inhibit the growth of Gram-positive accompanying bacterial flora. Di-potassium hydrogen phosphate is the buffering agent to stabilize the pH.mTSB is made selective for E. coli O157:H7 by adding bile salts in the media along with Novobiocin supplementation.

Application

mTSB Broth with Novobiocin is recommended for the selective enrichment of enterohemorrhagic E. coli (EHEC) in foods, especially meat and poultry products.The medium meets the requirements of the DIN Norm 10167 for the detection of E. coli serotype O157:H7 in foods. It is also recommended by the FDA-BAM as a method for the isolation of enterohemorrhagic E. coli (EHEC).

Preparation Note

Suspend 33 g in 1 litre of distilled water, autoclave at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isolation and molecular characterization of Salmonella enterica,
Escherichia coli O157:H7 and Shigella spp. from meat and dairy products
in Egypt
Ashraf M. Ahmed and Tadashi Shimamoto
International Journal of Food Microbiology, 57-62 (2014)
Optimization of enrichment and plating procedures for the recovery of Escherichia coli O111 and O26 from minced beef
T.M.G. Catarame
Journal of Applied Microbiology, 95(5) (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service