Skip to Content
Merck
All Photos(3)

Key Documents

103686BC

Millipore

mEHEC® Broth

Dehydrated culture medium for selective enrichment of pathogenic bacteria in selected foods

Synonym(s):

Modified Enterohemorrhagic Escherichia coli Broth

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$55.10
10 KG
$915.00

$55.10


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$55.10
10 KG
$915.00

About This Item

UNSPSC Code:
41106225
NACRES:
NA.74

$55.10


Estimated to ship onMarch 28, 2025


Request a Bulk Order

Quality Level

form

solid

shelf life

6‑36 mo.

packaging

pkg of (0.5 kg and 10 kg)

pH

7.0 ±0.2 (25 °C)

application(s)

food and beverages
microbiology

storage temp.

room temp

suitability

Escherichia coli H7 STEC
Escherichia coli non-O157
Escherichia coli (Enterohemorrhagic)
Salmonella sp

General description

mEHEC® Broth is a dehydrated granular culture medium used for selective enrichment of E. coli O157:H7, E. coli non-O157:H7 STEC, and Salmonella spp. in select food samples. The media contains tryptone that provides nitrogenous and carbonaceous compounds, long chain amino acids and other essential growth nutrients. Lactose is a fermentable sugar. Bile salts mixture inhibits gram-positive bacteria, especially bacilli and fecal Streptococci. Phosphates control the pH during the fermentation of lactose. Gas production in a fermentation tube within 24 hours or less is presumptive evidence of the presence of coliform bacteria.

Application

Dehydrated culture medium for selective enrichment of pathogenic bacteria in selected food and is intended for use with Assurance® GDS, TRANSIA® Plate, TRANSIA® AG, and VIP Gold.

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
TRANSIA is a registered trademark of Merck KGaA, Darmstadt, Germany
mEHEC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service