Skip to Content
Merck
All Photos(1)

Key Documents

74451

Supelco

Amberlite MB20 Ion Exchange Resin

mixed bed, >50 mesh, regenerated

Synonym(s):

mix mode ion exchange resin

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$130.00
1 KG
$414.00

$130.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$130.00
1 KG
$414.00

About This Item

MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

$130.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Product Name

Amberlite MB-20, mixed bed ion exchange resin, regenerated

form

beads

Quality Level

quality

mixed bed ion exchange resin
regenerated

technique(s)

LPLC: suitable

loss

~60% loss on drying, 110°C

matrix

PS-DVB

particle size

20-50 mesh

separation technique

mixed mode

General description

Amberlite MB-20 may be used for the removal of ions from aqueous solutions. Uses include primary water chemistry control in once-through systems, ultrapure water production, condensate polishing, in-process and column demineralization, and removal of ionic detergents (such as SDS) from protein samples.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. This resin can be used for the desalting of samples in carbohydrate determinations by HPLC using polymeric columns such as the Rezex type? Does it retain sugars?

    1 answer
    1. This resin is primarily used for water demineralization and deionization processes. Information regarding its affinity for carbohydrates is limited. For assistance with detailed method development, please contact local Technical Service.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service