Skip to Content
Merck
All Photos(3)

Documents

D2572

Sigma-Aldrich

AmberLite MB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511
NACRES:
NA.56

form

beads

technique(s)

ultrafiltration: suitable

matrix

Styrene-DVB, gel

matrix active group

functional group

particle size

<300 μm

pH

7

functional group

sulfonic acid (with ammonium ion)

storage temp.

room temp

General description

AmberLite MB mixed ion exchange resin is a ready-to-use mixed resin for the production of spot-free water, using a 1:1 equivalent mixture of strong acid cation and strong base anion exchange resin.

Application

AmberLite MB Mixed Ion Exchange Resin has been used for cleaning the nanoparticle dispersions. It has also been used in the purification of the cationic polystyrene (PS) seed latex.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Choosing mineral flotation collectors from large nanoparticle libraries
Abarca C, et al.
Journal of Colloid and Interface Science, 516, 423-430 (2018)
Relating nanoparticle shape and adhesiveness to performance as flotation collectors
Dong X, et al.
Industrial & Engineering Chemistry Research, 55(36), 9633-9638 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service