Skip to Content
Merck
All Photos(1)

Key Documents

73009

Millipore

ECC ChromoSelect Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

E. coli Coliform Agar ChromoSelect, ECC Agar ChromoSelect

Sign Into View Organizational & Contract Pricing

Select a Size

56 G
$156.00

$156.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

Select a Size

Change View
56 G
$156.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

$156.00


Estimated to ship onMay 27, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15 g/L
chromogenic mixture, 20.3 g/L
disodium hydrogen phosphate, 3.5 g/L
lactose, 2.5 g/L
monopotassium hydrogen phosphate, 1.5 g/L
neutral red, 0.03 g/L
peptone, special, 5 g/L
sodium chloride, 5 g/L
yeast extract, 3 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

6.8±0.2 (25 °C)

application(s)

agriculture
bioburden testing
environmental
food and beverages
water monitoring

microbiology

suitability

nonselective and differential for Enterobacter spp.
nonselective and differential for Escherichia coli
nonselective and differential for Klebsiella spp.
nonselective and differential for coliforms

Application

ECC ChromoSelect Agar is a differential medium recommended for the presumptive identification of Escherichia coli and other coliforms in food and environmental samples. The chromogenic mixture containstwo chromogens as X-glucuronide and Salmon-GAL. X-glucuronide is cleaved by the enzyme beta glucuronidase produced by Escherichia coli. Salmon-GAL is cleaved by the enzyme galactosidase produced by majority of coliforns, including Escherichia coli. Color of E. coli colonies: blue/purple

Preparation Note

Suspend 55.8 g in 1 litre distilled water. Sterilize by autoclaving at 121°C for 15 minutes. Cool to 50°C and pour into sterile petri plates

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S A Omolajaiye et al.
BioMed research international, 2020, 4250165-4250165 (2020-12-10)
Diarrhea has been reported as the leading cause of childhood mortality and morbidity globally but with disproportionate impacts in developing nations. Among bacterial etiologic agents of diarrhea, diarrheagenic Escherichia coli is the main cause of the disease among children under

Articles

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Questions

  1. What USP or AOAC method does the ECC agar comply with?

    1 answer
    1. This product is not tested according to USP or AOAC methods.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service