Skip to Content
Merck
All Photos(1)

Key Documents

39734

Millipore

Membrane Lactose Glucuronide Agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

MLGA, m-Lauryl Sulfate Chromogen Agar

Sign Into View Organizational & Contract Pricing

Select a Size

88 G
$163.00
500 G
$643.00

$163.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
88 G
$163.00
500 G
$643.00

About This Item

UNSPSC Code:
41171606
NACRES:
NA.85

$163.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 10 g/L
X-glucuronide (BCIG), 0.2 g/L
lactose, 30 g/L
peptone, 40 g/L
phenol red, 0.2 g/L
sodium lauryl sulfate, 1.0 g/L
sodium pyruvate, 0.5 g/L
yeast extract, 6 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

agriculture
bioburden testing
clinical testing
environmental
food and beverages
water monitoring

microbiology

suitability

selective and differential for Enterobacter spp.
selective and differential for Escherichia coli
selective and differential for Klebsiella spp.
selective and differential for coliforms
selective and differential for enterovirulent E. coli (EEC)

Application

A chromogenic medium for the differentiation and enumeration of Escherichia coli and other coliforms, which simplifies the membrane filtration technique by reducing the number of filtration stages and the need for further confirmation steps. This medium is described in the Environment Agency′s report of UK, ′Methods for Examination of Waters and Associated Material - The Microbiology of Drinking Water (2002) - Part 4 - Methods for the isolation and enumeration of coliform bacteria and Escherichia coli (including E. coli O157:H7).

Preparation Note

Suspend 88g in 1 litre of distilled water. Mix well and sterilize by autoclaving at 121°C for 15 minutes. Cool the medium to 50°C and pour into sterile Petri dishes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

An article concerning selective growth media for differentiation and detection of Escherichia coli and other coliforms.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service