Skip to Content
Merck
All Photos(1)

Documents

72018

Supelco

Sodium tetraphenylborate

Selectophore, ≥99.5%

Synonym(s):

Tetraphenylboron sodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)4BNa
CAS Number:
Molecular Weight:
342.22
Beilstein:
3599783
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥99.5% (NT)
≥99.5%

form

powder

storage temp.

2-8°C

SMILES string

[Na+].c1ccc(cc1)[B-](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20B.Na/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24;/h1-20H;/q-1;+1

InChI key

HFSRCEJMTLMDLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium tetraphenylborate is categorized under the tetraalkylborate group of compounds. Visit our Sensor Applications portal to learn more.

Application

Sodium tetraphenylborate (NaTPB) may be used as an anion discriminator in the poly(vinyl chloride) (PVC) based membrane potentiometric sensor for the determination of copper in aqueous solutions. Its addition to membranes has been proven to be beneficial towards optimizing sensing selectivity and decreasing bulk membrane impedance which result in substantial improvement of Cu2+ -selective potentiometric sensor performance. NaTBP has also been investigated as a lipophilic additive in terbium(III) membrane electrode for its potential response towards terbium(III) ions. It may be used as an aqueous derivatization reagent for organomercury prior to its determination in environmental samples by gas chromatography (GC) combined with atomic fluorescence spectrometry (AFS), microwave-induced plasma atomic emission spectrometry (AES), and mass spectrometry (MS).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of ethambutol-copper (II) complex and fabrication of PVC based membrane potentiometric sensor for copper.
Gupta VK, et al.
Talanta, 60(1), 149-160 (2003)
Construction of a novel potentiometric terbium (III) membrane sensor and its application for the determination of terbium ion in binary mixture and fluoride ion in a mouth wash preparation.
Zamani HA, et al.
Journal of the Brazilian Chemical Society, 17(7), 1297-1303 (2006)
Copper (II)-selective potentiometric sensors based on porphyrins in PVC matrix,.
Gupta VK, et al.
Sensors and Actuators B, Chemical, 117(1), 99-106 (2006)
Y Cai et al.
Journal of chromatography. A, 876(1-2), 147-155 (2000-05-24)
Several hyphenated analytical techniques, including gas chromatography (GC) coupled with atomic fluorescence spectrometry (AFS), microwave-induced plasma atomic emission spectrometry (AES), and mass spectrometry (MS), have been evaluated for methylmercury and ethylmercury analysis following aqueous derivatization with both sodium tetraethylborate and
A comparison of alkyl derivatization methods for speciation of mercury based on solid phase microextraction gas chromatography with furnace atomization plasma emission spectrometry detection.
Grinberg P, et al.
Journal of Analytical Atomic Spectrometry, 18(8), 902-909 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service