71853-M
Sodium perchlorate monohydrate
puriss. p.a., ACS reagent, ≥98.0% (T)
About This Item
Recommended Products
grade
ACS reagent
puriss. p.a.
Assay
≥98.0% (T)
form
crystalline
impurities
≤0.0005% total nitrogen (N)
pH
6.0-8.0 (25 °C, 50 mg/mL in H2O)
anion traces
chloride (Cl-): ≤30 mg/kg
sulfate (SO42-): ≤20 mg/kg
cation traces
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤3 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)
SMILES string
O.[Na+].[O-]Cl(=O)(=O)=O
InChI
1S/ClHO4.Na.H2O/c2-1(3,4)5;;/h(H,2,3,4,5);;1H2/q;+1;/p-1
InChI key
IXGNPUSUVRTQGW-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
Warning
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 1
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service