Skip to Content
Merck
All Photos(3)

Key Documents

71022ATMBC

Millipore

Assurance® GDS Salmonella HET Tq Kit for Automation

suitable for PCR testing in Food and Beverages applications

Sign Into View Organizational & Contract Pricing

Select a Size

576 TESTS
$14,270.00

$14,270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
576 TESTS
$14,270.00

About This Item

UNSPSC Code:
41106302
NACRES:
NA.85

$14,270.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Product Name

GDS Salmonella HET Tq Automation Kit, BioControl, Molecular based PCR test for the detection of Salmonellla spp., Salmonella Heidelberg, Salmonella Enteritidis, and Salmonella typhimurium in raw poultry and environmental samples

Agency

PCR, Immunomagnetic Separation (IMS)
AOAC® (PTM)

Quality Level

shelf life

6‑12 mo.

packaging

pkg of 576 tests

manufacturer/tradename

Assurance GDS®
BioControl Systems 71022-576ATM

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

BioControl GDS
for use with GDS Rotor-Gene

storage temp.

2-8°C

suitability

Bacillus cereus (NHE)
Salmonella spp.

Related Categories

General description

GDS, genetic detection system, for Salmonella HET Tq is an automated nucleic acid amplification system for the detection of Salmonella spp., S. Heidelberg, S. Enteritidis and/or S. Typhimurium (HET) in environmental boot swabs, poultry rinses, raw poultry and raw ground poultry.

Components

Each Assurance GDS for Salmonella HET Tq 100 kit contains the following:
  • Salmonella HET Amplification Tubes Tq (0.2 mL)
  • Salmonella HET Concentration Reagent

The following are also necessary and should be ordered separately:
  • 61031BC Wash Solution Kit
  • 34724BC Resuspension Buffer Tq

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

AOAC is a registered trademark of AOAC International
Assurance GDS is a registered trademark of BioControl Systems, Inc.

required but not provided

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service