Skip to Content
Merck
All Photos(1)

Key Documents

73044BC

Millipore

GDS Sample Wells

BioControl, For use with GDS Sample Wells Base

Synonym(s):

Sample Wells for PCR Sample Prep

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104017
Pricing and availability is not currently available.

manufacturer/tradename

BioControl Systems 73044

packaging

pkg of (Bag of 200)

storage condition

room temperature

technique(s)

PCR: suitable

application(s)

food and beverages
pathogen testing

compatibility

for use with GDS Rotor-Gene

storage temp.

room temp

General description

GDS Sample Wells are the disposables required to perform Assurance® GDS assays. Food sample is added to the sample wells, along with a Concentration Reagent (IMS beads) as part of the initial PCR sample preparation for GDS. Adhesive film strips are used to seal the sample wells.

Application

GDS Sample Wells are the disposables used during PCR sample preparation for Assurance® GDS assays. The sample wells are to be used with the GDS Sample Wells Base.

Components

Bag of 200

Testing Method

PCR, Immunomagnetic Separation (IMS)

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany

also commonly purchased with this product

Product No.
Description
Pricing

required but not provided


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service