Skip to Content
Merck
All Photos(1)

Key Documents

70158

Supelco

(−)-Myrtenol

analytical standard

Synonym(s):

(1R)-2-Pinen-10-ol, (1R)-6,6-Dimethylbicyclo[3.1.1]hept-2-ene-2-methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Beilstein:
2206583
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (sum of enantiomers, GC)

optical activity

[α]/D -56±1°, c = 1 in chloroform

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.496

bp

220-222 °C (lit.)

density

0.981 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)[C@H]2CC=C(CO)[C@@H]1C2

InChI

1S/C10H16O/c1-10(2)8-4-3-7(6-11)9(10)5-8/h3,8-9,11H,4-6H2,1-2H3/t8-,9-/m0/s1

InChI key

RXBQNMWIQKOSCS-IUCAKERBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(−)-Myrtenol is a naturally occurring monoterpene alcohol, belonging to the class of volatile organic compounds. It is widely used in fragrance, cosmetic products, cleaning agents and flavoring agents in a variety of food products.[1]

Application

(−)-Myrtenol may be used as an analytical reference standard for the quantification of the analyte in biological samples using high-performance liquid chromatography with fluorescence detection and electrospray ionization-mass spectrometry.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Use as chiral building block[2][3][4][5]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensitive determination of monoterpene alcohols in urine by HPLC-FLD combined with ESI-MS detection after online-solid phase extraction of the monoterpene-coumarincarbamate derivates
Meesters.W.JR, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 875(2), 444-450 (2008)
L. Borowiecki et al.
Liebigs Ann. Chem., 1775-1775 (1982)
R. Pellegata et al.
Helvetica Chimica Acta, 70, 71-71 (1987)
Synthesis of stable thromboxane A2 analogs: pinane thromboxane A2 (PTA2) and carbocyclic thromboxane A2 (CTA2).
K C Nicolaou et al.
Methods in enzymology, 86, 400-409 (1982-01-01)
Y. Bounameaux et al.
Helvetica Chimica Acta, 66, 989-989 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service