Skip to Content
Merck
All Photos(1)

Key Documents

192384

Sigma-Aldrich

(−)-Carveol, mixture of isomers

97%

Synonym(s):

p-Mentha-6,8-dien-2-ol

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$101.00

$101.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$101.00

About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

$101.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

optical activity

[α]20/D −112°, c = 1 in chloroform

refractive index

n20/D 1.496 (lit.)

bp

226-227 °C/751 mmHg (lit.)

density

0.958 g/mL at 25 °C (lit.)

functional group

hydroxyl

SMILES string

CC(=C)[C@@H]1CC=C(C)C(O)C1

InChI

1S/C10H16O/c1-7(2)9-5-4-8(3)10(11)6-9/h4,9-11H,1,5-6H2,2-3H3/t9-,10?/m1/s1

InChI key

BAVONGHXFVOKBV-YHMJZVADSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imran Malik et al.
Frontiers in neuroscience, 14, 659-659 (2020-07-28)
Ischemic stroke is a severe neurological disorder with a high prevalence rate in developed countries. It is characterized by permanent or transient cerebral ischemia and it activates syndrome of pathological events such as membrane depolarization, glutamate excitotoxicity, and intracellular calcium
M Wüst et al.
Archives of biochemistry and biophysics, 387(1), 125-136 (2001-05-23)
Limonene enantiomers and substrate analogs, including specifically fluorinated derivatives, were utilized to probe active site interactions with recombinant (-)-(4S)-limonene-3-hydroxylase (CYP71D13) and (-)-(4S)-limonene-6-hydroxylase (CYP71D18) from mint (Mentha) species. (-)-(4S)-Limonene is hydroxylated by both enzymes at the designated C3- and C6-allylic positions
W A Duetz et al.
Applied and environmental microbiology, 67(6), 2829-2832 (2001-05-26)
The toluene-degrading strain Rhodococcus opacus PWD4 was found to hydroxylate D-limonene exclusively in the 6-position, yielding enantiomerically pure (+) trans-carveol and traces of (+) carvone. This biotransformation was studied using cells cultivated in chemostat culture with toluene as a carbon
Jenna L E Morrish et al.
Biotechnology and bioengineering, 101(4), 768-775 (2008-05-15)
The microbial biotransformation of (-)-trans-carveol to the flavor and fragrance compound (R)-(-)-carvone by Rhodococcus erythropolis DCL14 was carried out in a 3 L two phase partitioning bioreactor with an immiscible liquid second phase in an effort to improve upon the
Jenna L E Morrish et al.
Biotechnology letters, 30(7), 1245-1250 (2008-03-11)
The aqueous substrate and product toxicity thresholds in the microbial biotransformation of (-)-trans-carveol to the fragrance/flavor compound (R)-(-)-carvone by Rhodococcus erythropolis were determined. Above aqueous phase concentrations of approx. 500 mg carveol/l and 200-600 mg carvone/l, the biotransformation activity of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service