Skip to Content
Merck
All Photos(1)

Key Documents

69729

Supelco

Elemental Impurities according to ICH Q3D oral, Standard 3

TraceCERT®, in nitric acid and hydrofluoric acid (5% nitric acid and <0.5% hydrofluoric acid ), applicable for testing acc. to USP<232>, Ph.Eur. Gen. Chapter 5.20

Synonym(s):

ICH Q3D oral elemental impurities

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$263.00

$263.00


In StockDetails


Request a Bulk Order

Select a Size

Change View
100 ML
$263.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$263.00


In StockDetails


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

Agency

according to ICH Q3D
according to Ph. Eur.
according to USP

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

composition

Ba, 140 mg/L
Cr, 1100 mg/L
Cu, 300 mg/L
Li, 55 mg/L
Mo, 300 mg/L
Sb, 120 mg/L
Sn, 600 mg/L

concentration

in nitric acid and hydrofluoric acid (5% nitric acid and <0.5% hydrofluoric acid )

technique(s)

ICP: suitable

format

multi-component solution

General description

This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at https://www.sigma-aldrich.com to view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Other Notes

We provide a diverse range of certified reference materials, including various elemental impurity mixes and single elements, with concentration limits that comply with ICH Q3D guidelines. For more information please visit our Inorganic Elemental Impurity Mix Standards.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service