Skip to Content
Merck
All Photos(1)

Key Documents

69710

Sigma-Aldrich

Methyl violet

for microscopy (Bact., Bot., Hist.), indicator (pH 0.1-2.0), mixture of polymethylated pararosaniline hydrochlorides

Synonym(s):

Methyl violet 2B, Basic Violet 1

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$44.80
100 G
$99.00
500 G
$366.00

$44.80


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$44.80
100 G
$99.00
500 G
$366.00

About This Item

CAS Number:
Colour Index Number:
42535
Beilstein:
8464808
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

$44.80


Estimated to ship onMay 28, 2025


Request a Bulk Order

grade

for microscopy (Bact., Bot., Hist.)
indicator (pH 0.1-2.0)

Quality Level

form

powder or crystals

color

green to very dark green

pH

1-2

mp

137 °C (dec.) (lit.)

solubility

ethanol and water: 0.01 g/10 mL

εmax

≥1500 at 579-589 nm in 50% ethanol

suitability

suitable for microscopy

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl.C\N=C1/C=C\C(C=C1)=C(/c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C

InChI

1S/C24H27N3.ClH/c1-25-21-12-6-18(7-13-21)24(19-8-14-22(15-9-19)26(2)3)20-10-16-23(17-11-20)27(4)5;/h6-17H,1-5H3;1H

InChI key

JFTBTTPUYRGXDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl Violet 2B is a mixture of polymethylated (4,5 and 6) pararosaniline chlorides
Methyl violet is a triphenylmethane dye. It is a mixture of tetra-, penta-, and hexa- methyl pararosaniline. [1]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

John D. Bancroft
Theory and Practice of Histological Techniques, 725-725 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service