Skip to Content
Merck
All Photos(2)

Key Documents

277339

Sigma-Aldrich

Methyl Purple

in H2O

Synonym(s):

Methyl Purple indicator solution

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$235.00

$235.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

Select a Size

Change View
1 L
$235.00

About This Item

Empirical Formula (Hill Notation):
C67H62N8Na4O14S3
CAS Number:
Molecular Weight:
1391.41
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$235.00


Estimated to ship onMay 30, 2025


Request a Bulk Order

form

liquid

Quality Level

concentration

in H2O

pH

4.8-5.4, violet to green

mp

0 °C

density

1.001 g/mL at 25 °C

λmax

409 nm
618 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1ccc(cc1)N=Nc2ccccc2C(=O)O[Na].CN(C)c3ccc(cc3)N=Nc4ccccc4C(=O)O[Na].CCN(Cc5cccc(c5)S(=O)(=O)O[Na])c6ccc(cc6)C(=C7/C=CC(C=C7)=[N+](CC)Cc8cccc(c8)S([O-])(=O)=O)\c9ccc(O)cc9S(=O)(=O)O[Na]

InChI

1S/C37H36N2O10S3.2C15H15N3O2.4Na/c1-3-38(24-26-7-5-9-33(21-26)50(41,42)43)30-15-11-28(12-16-30)37(35-20-19-32(40)23-36(35)52(47,48)49)29-13-17-31(18-14-29)39(4-2)25-27-8-6-10-34(22-27)51(44,45)46;2*1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20;;;;/h5-23H,3-4,24-25H2,1-2H3,(H3,41,42,43,44,45,46,47,48,49);2*3-10H,1-2H3,(H,19,20);;;;/q;;;4*+1/p-4/b;2*17-16+;;;;

InChI key

CNJVTYXLOYTWPD-HFZRWZJVSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Methyl purple is a quinone-imide redox dye.

Biochem/physiol Actions

Methyl Purple is a green colored powder. It changes color from purple (pH 4.8) to green (pH 5.4). Methyl Purple acts as an antimicrobial agent. In addition, it also aids in detection of lactic acid bacteria and determination of iron in human hair.[1]

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Graan et al.
Analytical biochemistry, 144(1), 193-198 (1985-01-01)
Methyl purple is a recently introduced quinone-imide redox dye that has been shown to be an exceptionally sensitive monitor of photosystem I activity in chloroplasts. This compound has a wide range of potential applications for studies of photosynthetic electron transfer
R. W. Sabnis
Handbook of Acid-Base Indicators (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service