Skip to Content
Merck
All Photos(2)

Key Documents

66008BC

Millipore

SimPlate® Total Coliform & E. coli CI

SimPlate® Total Coliform & E. coli CI
1 of 1 reviewers received a sample product or took part in a promotion

BioControl, Quantitative results for Coliforms and E. coli from food and environmental samples in just 24 hours

Synonym(s):

CEC-CI (Color Indicator)

Sign Into View Organizational & Contract Pricing

Select a Size

100 TEST
$297.00
500 TEST
$977.00

$297.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
100 TEST
$297.00
500 TEST
$977.00

About This Item

UNSPSC Code:
41106200
NACRES:
NA.85
material:
plastic vial (containing dehydrated culture media)
suitability:
coliforms: Escherichia coli
feature:
fluorescent indicator

$297.00


Estimated to ship onMay 25, 2025


Request a Bulk Order

material

plastic vial (containing dehydrated culture media)

size

100 tests
500 tests

Agency

AOAC® OMA

form

dry powder

shelf life

6‑18 mo.

feature

fluorescent indicator

packaging

container of 100

manufacturer/tradename

BioControl Systems 66008-100
BioControl Systems 66008-500

storage condition

dry media

application(s)

food and beverages

compatibility

for use with SimPlate (65009BC)

storage temp.

room temp

suitability

coliforms: Escherichia coli

General description

SimPlate for Total Coliform & E. coli Color Indicator (CEc-CI) method is used for the detection and quantification of the Total Coliform and E. coli populations. It is based on Binary Detection Technology (BDT) which equates the presence of Total Coliform to the presence of a color change in a medium and E. coli is detected by the presence of a blue fluorescent color change in the medium due to the action of the enzyme and Β-glucuronidase. The medium/sample mixture is dispensed into a SimPlate device, and incubated for 24-28 h. The Total Coliform and E. coli count is determined by counting the wells with changed color and referring to the SimPlate Conversion Table. Colored wells indicate the presence of Total Coliforms, and colored wells that fluoresce indicate the presence of E. coli. The SimPlate device is packaged separately.

Application

AOAC Official Method 2005.03

Features and Benefits

  • Total Coliform and E. coli results are available from a single plate. Proprietary chromogenic media and patented plating device with isolation wells combine to produce easily counted results. Because results are not dependent on gas formation, results are more easily interpreted than tube or film-based methods.
  • While other plating methods require duplicate plating of samples, and the standard MPN method requires multiple dilutions and at least nine tubes of media, SimPlate has been validated to provide equivalent results with just a single plate. Combined with the reduction of necessary sample dilutions, made possible by the broad counting range of the plate (1-738), SimPlate reduces the volume of media and number of tests required for each sample, helping to reduce overall testing costs.

Components

2 packages of 50 single-test vials (66008-100), or 1 package of 50 multi-test (10 tests/vial) vials (66008-500)

Testing Method

Binary Detection Technology®

Other Notes

Required for use: SimPlate Device (65009BC)

Legal Information

AOAC is a registered trademark of AOAC International
Binary Detection Technology is a registered trademark of Merck KGaA, Darmstadt, Germany
SimPlate is a registered trademark of Merck KGaA, Darmstadt, Germany

Kit Components Also Available Separately

Product No.
Description
SDS

  • SimPlate Coliform & E. coli-Color Indicator (CEc-CI) Single Dose Fill, 100 test

  • SimPlate Coliform & E. coli-Color Indicator (CEc-CI) Multi-Dose Fill, 500 test

also commonly purchased with this product

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is this product EPA approved? 66

    1 answer
    1. SimPlate® Total Coliform & E. coli CI, Cat # 66008 is AOAC official method not EP.

      Helpful?

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Los Fresnos Tx, 78566
    • Review 1
    • Votes 0
    5 out of 5 stars.

    Great

    i really love this material and makes our testing easier

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service