Skip to Content
Merck
All Photos(1)

Key Documents

63012BC

Millipore

Supplement R, SimPlate®

BioControl, For use with SimPlate Coliform & E. coli Color Indicator

Synonym(s):

Supplement

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
$62.10

$62.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EACH
$62.10

About This Item

UNSPSC Code:
41106200
NACRES:
NA.85

$62.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

material

plastic vial (containing dehydrated culture media)

form

liquid

shelf life

6‑15 mo.

packaging

bottle of 1 20 mL

manufacturer/tradename

BioControl Systems 63012-100

storage condition

protect from light

application(s)

food and beverages

compatibility

for use with SimPlate (66008BC)

storage temp.

2-8°C

suitability

coliforms: Escherichia coli

Related Categories

General description

Supplement R is an additional reagent for use in the SimPlate® Coliform & E. coli. SimPlate® Binary Detection Technology combines proprietary media and a patented plating device to provide fast and accurate quantitative results for key groups of indicator organisms. Defined substrates incorporated into the media are utilized to detect multiple enzymes and redox reactions associated with the target organisms resulting in chromogenic or fluorescent changes to the media, providing easy-to-read results. Supplement R is added to aid the recovery of coliforms from high microbial samples.

Application

Supplement R is added to SimPlate® trays to recover the coliforms from high microbial samples for the detection in foods, ingredients, and environmental samples without the need for optimization of reaction and cycle conditions.

Components

1 bottle, approx. 20 mL (adequate for 200 tests)

Other Notes

For use with SimPlate Total Coliform & E. coli CI (66008BC)

Legal Information

SimPlate is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service