Skip to Content
Merck
All Photos(1)

Key Documents

648531

Sigma-Aldrich

N,N-Dimethylformamide

suitable for HPLC, ≥99.9%

Synonym(s):

DMF, NSC 5356

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCON(CH3)2
CAS Number:
Molecular Weight:
73.09
Beilstein:
605365
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
food and beverages
bp:
153 °C (lit.)
vapor pressure:
2.7 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

HPLC grade

Quality Level

vapor density

2.5 (vs air)

vapor pressure

2.7 mmHg ( 20 °C)

Assay

≥99.9%

form

liquid

autoignition temp.

833 °F

expl. lim.

15.2 %

technique(s)

HPLC: suitable

impurities

≤0.030% water

evapn. residue

≤0.0005%

refractive index

n20/D 1.430 (lit.)

bp

153 °C (lit.)

mp

−61 °C (lit.)

density

0.944 g/mL (lit.)

absorption

0.010 at 340 nm
0.010 at 400 nm
0.05 at 310 nm
0.10 at 295 nm
0.30 at 275 nm
1.00 at 270 nm

application(s)

food and beverages

SMILES string

[H]C(=O)N(C)C

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethylformamide (DMF) is the commonly employed solvent for chemical reactions. DMF is a useful solvent employed for the isolation of chlorophyll from plant tissues.[1] It is widely employed reagent in organic synthesis. It plays multiple roles in various reactions such as solvent, dehydrating agent, reducing agent as well as catalyst. It is a multipurpose building block for the synthesis of compounds containing O, -CO, -NMe2, -CONMe2, -Me, -CHO as functional groups.[2] Dielectric relaxation experiments on DMF have been conducted between 238.15K and 338.15K.[3] The thermophysical properties of the molecular interactions between DMF and the ionic liquids, ammonium salts and imidazolium salts have been studied.[4] The characteristics of solute-solvent interactions between ammonium nitrate and DMF have been investigated.[5]

Application

Solvent for many hydrophobic organic compounds.

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

135.5 °F - closed cup

Flash Point(C)

57.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volumetric properties of ammonium nitrate in N,N-dimethylformamide.
Vranes M, et al.
The Journal of Chemical Thermodynamics, 54, 245-249 (2012)
Measurements and molecular interactions for N,N-dimethylformamide with ionic liquid mixed solvents.
Attri P, et al.
The Journal of Physical Chemistry B, 114(18), 6126-6133 (2010)
Synthesis and electrochemical characterization of aPEO-based polymer electrolytes.
Rodrigues LC, et al.
Journal of Solid State Electrochemistry, 16(4), 1623-1629 (2012)
The dynamics of liquid formamide, N-methylformamide, N,N-dimethylformamide, and N,N-dimethylacetamide. A dielectric relaxation study.
Barthel J, et al.
Journal of Molecular Liquids, 98, 51-69 (2002)
W P Inskeep et al.
Plant physiology, 77(2), 483-485 (1985-02-01)
We found inconsistencies in the commonly used data for chlorophyll analysis in 80% acetone. Recently developed extinction coefficients for chlorophyll b in N,N-dimethylformamide (DMF) based on values from 80% acetone are low as a result of these inconsistencies. We determined

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service