Skip to Content
Merck
All Photos(1)

Key Documents

1601500

USP

Residual Solvent Class 2 - N,N-Dimethylformamide

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N,N-Dimethylformamide solution

Sign Into View Organizational & Contract Pricing

Select a Size

3 X 1.2 ML
$573.00

$573.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
3 X 1.2 ML
$573.00

About This Item

Empirical Formula (Hill Notation):
C3H7NO
CAS Number:
Molecular Weight:
73.09
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$573.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N(C)(C)C=O

InChI

1S/C3H7NO/c1-4(2)3-5/h3H,1-2H3

InChI key

ZMXDDKWLCZADIW-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Residual Solvent Class 2 - N,N-Dimethylformamide USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cui Wang et al.
Environmental pollution (Barking, Essex : 1987), 190, 133-138 (2014-04-22)
Occupational and experimental studies have revealed the organs most affected by dimethylformamide (DMF) are liver and gastrointestinal tract. However, few studies have focused on the potential effect of outdoor pollution of DMF. This study examined the health risk of hospitalization
K Tanaka et al.
Sangyo igaku. Japanese journal of industrial health, 24(1), 3-12 (1982-01-01)
Industrial use of N, N-dimethylformamide (DMF) is increasing in recent years. The toxic properties of this substance are reviewed from the standpoint of industrial hygiene. Hepatotoxicity has been emphasized by many authors. Care should be taken against the skin absorption
Shengtao Ding et al.
Angewandte Chemie (International ed. in English), 51(37), 9226-9237 (2012-08-30)
Often used as a common solvent for chemical reations and utilized widely in industry as a reagent, N,N-dimethylformamide (DMF) has played an important role in organic synthesis for a long time. Numerous highly useful articles and reviews discussing its utilizations
Metabolism of N,N-dimethylformamide: key to the understanding of its toxicity.
A Gescher
Chemical research in toxicology, 6(3), 245-251 (1993-05-01)
Xiaowen Xu et al.
Organic letters, 16(13), 3540-3543 (2014-06-17)
A novel straightforward synthesis of both symmetrical and unsymmetrical 2,4-disubstituted-1,3,5-triazines via aerobic copper-catalyzed cyclization of amidines with DMF as a one-carbon synthon has been developed. The presented method allows synthesizing the products that are currently inaccessible or challenging to prepare

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service