Skip to Content
Merck
All Photos(1)

Key Documents

63080

Sigma-Aldrich

Magnesium phosphate dibasic trihydrate

BioUltra, ≥98.0% (KT)

Synonym(s):

Magnesium hydrogen phosphate trihydrate, Newberyite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgHPO4 · 3H2O
CAS Number:
Molecular Weight:
174.33
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Assay:
≥98.0% (KT)
solubility:
5 M HCl: 0.1 M at 20 °C, clear, colorless

product line

BioUltra

Quality Level

Assay

≥98.0% (KT)

impurities

insoluble matter, passes filter test

loss

~34% loss on ignition, 800 °C

solubility

5 M HCl: 0.1 M at 20 °C, clear, colorless

density

2.123 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Al: ≤100 mg/kg
As: ≤0.2 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤2000 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤500 mg/kg
Ni: ≤50 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in 5 M HCl

UV absorption

λ: 260 nm Amax: 0.035
λ: 280 nm Amax: 0.035

SMILES string

[Mg++].[H]O[H].[H]O[H].[H]O[H].OP([O-])([O-])=O

InChI

1S/Mg.H3O4P.3H2O/c;1-5(2,3)4;;;/h;(H3,1,2,3,4);3*1H2/q+2;;;;/p-2

InChI key

OKIWLDVQGKRUNR-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium hydrogen phosphate trihydrate (Newberyite), a biodegradable cave mineral, may be used to study its physical and reactive chemistries.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vesna Babić-Ivancić et al.
Water research, 40(18), 3447-3455 (2006-09-19)
The influence of the initial reactant concentrations on the composition of the solid phases formed in the precipitation system MgCl(2)-NH(4)H(2)PO(4)-NaOH-H(2)O was investigated. The precipitation diagram constructed shows the approximate concentration regions within which struvite, newberyite, and their mixtures exist at
Raman spectroscopy of newberyite Mg(PO3OH)?3H2O: a cave mineral.
Frost RL, Palmer SJ, Pogson RE.
Spectrochimica Acta Part A: Molecular Spectroscopy, 79, 1149-1153 (2011)
Jasminka Kontrec et al.
Collegium antropologicum, 29(1), 289-294 (2005-08-25)
The influence of the initial reactant concentrations (c(i)(Mg)tot = 5.0 x 10(6) to 5.0 x 10(-1) mol dm(-3), c(i)(P)tot = c(i)(NH4)tot = 1.0 x 10(-3) to 5.0 x 10(-1) mol dm(-3)) and temperature (25 and 37 degrees C) on the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service