Skip to Content
Merck
All Photos(1)

Key Documents

59603

Supelco

Terbutryn-(S-methyl-d3)

PESTANAL®, analytical standard

Synonym(s):

N-tert-Butyl-N′-ethyl-6-methyl-d3-thio-1,3,5-triazine-2,4-diamine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502

grade

analytical standard

Quality Level

isotopic purity

≥98.5 atom % D

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service