Skip to Content
Merck
All Photos(1)

Key Documents

59320

Sigma-Aldrich

Isopropylamine

puriss., ≥99.5% (GC)

Synonym(s):

2-Aminopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNH2
CAS Number:
Molecular Weight:
59.11
Beilstein:
605259
EC Number:
MDL number:
UNSPSC Code:
12352100

vapor density

2.04 (vs air)

vapor pressure

9.2 psi ( 20 °C)

grade

puriss.

Assay

≥99.5% (GC)

autoignition temp.

755 °F

expl. lim.

10.4 %

impurities

≤0.5% water

refractive index

n20/D 1.374 (lit.)
n20/D 1.374

bp

33-34 °C (lit.)

density

0.688 g/mL at 20 °C (lit.)

SMILES string

CC(C)N

InChI

1S/C3H9N/c1-3(2)4/h3H,4H2,1-2H3

InChI key

JJWLVOIRVHMVIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service