Skip to Content
Merck
All Photos(2)

Key Documents

57981

Supelco

5-Nonadecylresorcinol

analytical standard

Synonym(s):

1,3-Dihydroxy-5-nonadecylbenzene, 5-Nonadecyl-1,3-benzenediol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H44O2
CAS Number:
Molecular Weight:
376.62
Beilstein:
2387022
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCCCCCCc1cc(O)cc(O)c1

InChI

1S/C25H44O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23-20-24(26)22-25(27)21-23/h20-22,26-27H,2-19H2,1H3

InChI key

PUNOCEUUYUXUGR-UHFFFAOYSA-N

General description

5-Nonadecylresorcinol is an alk(en)yl resorcinol belonging to the class of amphiphilic phenolic lipids, generally found in wheat, spelt, barley, rye, peas, ginkgo, mango peal/pulp, etc. It exhibits pharmacological properties such as anti-inflammatory, anti-carcinogenic and anti-microbial activities.

Application

5-Nonadecylresorcinol may be used as an analytical reference standard for the quantification of the analyte in fruits and rye grain using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatographic analysis of alkylresorcinols in rye (Secale cereale L) grains
Ross BA, et al.
Journal of the Science of Food and Agriculture, 81(14), 1405-1411 (2001)
A Kozubek et al.
Biochimica et biophysica acta, 944(3), 465-472 (1988-10-20)
The effect of alk(en)ylresorcinol homologs (5-(n-nonadecyl)- and 5-(n-nonadecenyl)resorcinol) on the mobility of 5-doxyl- and 12-doxylstearate spin probes incorporated into DMPC, DMPC-cholesterol and erythrocyte membranes was studied. It was found that both homologs affect the properties of hydrophobic environment of the
Jose Rodríguez-Morató et al.
Rapid communications in mass spectrometry : RCM, 34(14), e8805-e8805 (2020-04-17)
Consumption of whole grains is negatively associated with cardiovascular disease (CVD) risk but quantification of whole-grain intake is challenging. Alkylresorcinols (ARs) are biomarkers of whole-grain intake. Current methods for AR quantification involve a time-consuming multi-step separation process that hampers applicability
Development and validation of an HPLC method for the determination of alk (en) ylresorcinols using rapid ultrasound-assisted extraction of mango peels and rye grains
Geerkens.HC, et al.
Food Chemistry, 169, 261-269 (2015)
Yingdong Zhu et al.
Bioorganic & medicinal chemistry, 19(13), 3973-3982 (2011-06-11)
We and others have found that wheat bran oil is the active constituent in wheat bran for colon cancer prevention. However, the active components in wheat bran oil are still unknown. Using human colon cancer cells (HCT-116 and HT-29) as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service