Skip to Content
Merck
All Photos(1)

Key Documents

57524

Supelco

1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran

analytical standard

Synonym(s):

1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylindeno[5,6-c]pyran, Hexamethylindanopyran

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$162.00

$162.00


Estimated to ship onMay 11, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$162.00

About This Item

Empirical Formula (Hill Notation):
C18H26O
CAS Number:
Molecular Weight:
258.40
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$162.00


Estimated to ship onMay 11, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥80.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.510-1.540

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC1(C)C(C)C(C)(C)C2=CC3=C(C(C)COC3)C=C21

InChI

1S/C18H26O/c1-11-14-10-16-15(9-13(14)7-8-19-11)17(3,4)12(2)18(16,5)6/h9-12H,7-8H2,1-6H3

InChI key

YWMTVUUYOAIDBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran may be used as a reference standard in the determination of the analyte in fragrances using bi-dimensional gas chromatography coupled with quadrupole mass spectrometry (GCxGC-QMS).[1]

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

291.2 °F - Pensky-Martens closed cup

Flash Point(C)

144 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Routine quantification of 54 allergens in fragrances using comprehensive two-dimensional gas chromatography-quadrupole mass spectrometry with dual parallel secondary columns. Part I: Method development.
Belhassen E, et al.
Flavour and Fragrance Journal, 33(1), 63-74 (2018)
Óscar Castro et al.
Journal of chromatography. A, 1591, 55-61 (2019-01-20)
A novel solid phase microextraction Arrow (SPME Arrow) system has been applied for the first time to determine synthetic musk fragrances in fish samples. The lack of regulation concerning the concentration of musk fragrances in fish along with the risk

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service