Skip to Content
Merck
All Photos(1)

Key Documents

75831

Supelco

Santalol, α- and β-isomers

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$185.00

$185.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$185.00

About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$185.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥80% (sum of isomers)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.507 (lit.)

bp

101-103 °C/0.1 mmHg (lit.)

density

0.968 g/mL at 25 °C (lit.)

format

neat

storage temp.

−20°C

SMILES string

OC\C(=C\CC[C@]1([C@@H]2C[C@@H](CC2)C1=C)C)\C

InChI

1S/C15H24O/c1-11(10-16)5-4-8-15(3)12(2)13-6-7-14(15)9-13/h5,13-14,16H,2,4,6-10H2,1,3H3/b11-5+/t13-,14+,15-/m1/s1

InChI key

OJYKYCDSGQGTRJ-INLOORNJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Santalol, α- and β-isomers are used as analytical standards for the quantification of the analytes in fragrances using flow-modulation comprehensive two-dimensional gas chromatography coupled to mass spectrometry (FM GCxGC/MS).

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Is this a racemic mixture? If not what is the percentage of alpha: beta santalols?

    1 answer
    1. This product is not a racemic mixture. The majority of this material is the alpha isomer. The isomeric content is lot specific and reported in the product Certificate of Analysis. However, only the percentage sum of isomers and confirmation of the presence of both alpha- and beta-santalol are reported. Please see the link below to review a sample or lot specific Certificate of Analysis:
      https://www.sigmaaldrich.com/US/en/product/sial/75831#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service