Skip to Content
Merck
All Photos(1)

Key Documents

56890

Supelco

Conductivity Standard

conductivity 300 μS/cm at 25 °C

Synonym(s):

Conductivity Calibration Solution

Sign Into View Organizational & Contract Pricing

Select a Size

500 ML
$319.00

$319.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

Select a Size

Change View
500 ML
$319.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

$319.00


Estimated to ship onApril 03, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

≤2.5% KCl (T)

conductivity

297.0-303.0 μS/cm at 25 °C

density

0.995-1.014 g/cm3 at 20 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

single component solution

Looking for similar products? Visit Product Comparison Guide

General description

Conductivity standard is used for the calibration of conductivity measurements.[1] The conductivity standard solution, which can be used for conductivity measurements can be sodium chloride or potassium chloride solution of a definite concentration.[2]

Application


  • Development of Conductivity Method as an Alternative to Titration for Hydrolytic Resistance Testing Used for Evaluation of Glass Vials Used in Pharmaceutical Industry.: This study highlights the use of conductivity calibration standards in the pharmaceutical industry for evaluating the hydrolytic resistance of glass vials. By employing high-precision conductivity measurements, the research demonstrates an efficient alternative to traditional titration methods, ensuring accurate and reliable testing for pharmaceutical applications (Fujimori et al., 2017).

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - STOT RE 2 Oral

Target Organs

Liver,Kidney

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bela G, et al.
Analysis and Analyzers (2016)
Volunteer stream monitoring a methods manual null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service