56481
Cellobiose Disks
suitable for microbiology, Sterile filter paper discs impregnated with cellobiose
Synonym(s):
Carbohydrate discs
About This Item
Recommended Products
Quality Level
sterility
sterile
product line
BioChemika
shelf life
limited shelf life, expiry date on the label
packaging
pkg of 10 × 25 discs
technique(s)
microbe id | utilization test: suitable
application(s)
clinical testing
environmental
food and beverages
pharmaceutical
microbiology
storage temp.
2-8°C
suitability
Citrobacter spp.
Enterobacter spp.
Klebsiella spp.
bacteria
General description
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Salmonella, with 2,300+ serotypes, causes half of food-borne illnesses, often from dairy, poultry, and eggs.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service