Skip to Content
Merck
All Photos(2)

Key Documents

55549

Supelco

14-Deoxy-11,12-didehydroandrographolide

analytical standard

Synonym(s):

3-[(1E)-2-[(1R,4aS,5R,6R,8aR)-Decahydro-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethenyl]- 2(5H)-furanone, ent-(3β,11E)-3,19-Dihydroxy-8(17),11,13-labdatrien-16,15-olide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$552.00

$552.00


Estimated to ship onApril 08, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$552.00

About This Item

Empirical Formula (Hill Notation):
C20H28O4
CAS Number:
Molecular Weight:
332.43
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$552.00


Estimated to ship onApril 08, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥95% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

C[C@@]1(CO)[C@H](O)CC[C@]2(C)[C@@H]1CCC(=C)[C@H]2\C=C\C3=CCOC3=O

InChI

1S/C20H28O4/c1-13-4-7-16-19(2,10-8-17(22)20(16,3)12-21)15(13)6-5-14-9-11-24-18(14)23/h5-6,9,15-17,21-22H,1,4,7-8,10-12H2,2-3H3/b6-5+/t15-,16+,17-,19+,20+/m1/s1

InChI key

XMJAJFVLHDIEHF-CRBRZBHVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

14-Deoxy-11,12-didehydroandrographolide is a major bitter diterpene lactone component, isolated from F82:F117Andrographis paniculata. It is commonly used for the treatment of fever and infectious fever-causing diseases.[1]

Application

14-Deoxy-11,12-didehydroandrographolide may be used as an analytical reference standard in plant materials and commercial products of Andrographis paniculata using high-performance liquid chromatography technique.[2]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HPLC and HPTLC densitometric determination of andrographolides and antioxidant potential of Andrographis paniculata
Akowuah.A.G, et al.
J. Food Compos. Anal., 19(2-3), 118-126 (2006)
HPLC?PDA determination of bioactive diterpenoids from plant materials and commercial products of Andrographis paniculata
Li W and Fitzloff F.J
Journal of Liquid Chromatography and Related Technologies, 27(15), 2407-2420 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service