Skip to Content
Merck
All Photos(1)

Key Documents

365645

Sigma-Aldrich

Andrographolide

98%

Synonym(s):

3-dehydroandrographolide, Andrographis, Andrographiside aglycon

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$69.50
500 MG
$263.00

$69.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$69.50
500 MG
$263.00

About This Item

Empirical Formula (Hill Notation):
C20H30O5
CAS Number:
Molecular Weight:
350.45
Beilstein:
42762
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$69.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

powder

optical activity

[α]20/D −126°, c = 1.5 in acetic acid

mp

229-232 °C (lit.)

functional group

ester
hydroxyl

SMILES string

C[C@@]1(CO)[C@H](O)CC[C@@]2(C)[C@H](C\C=C3/[C@H](O)COC3=O)C(=C)CC[C@H]12

InChI

1S/C20H30O5/c1-12-4-7-16-19(2,9-8-17(23)20(16,3)11-21)14(12)6-5-13-15(22)10-25-18(13)24/h5,14-17,21-23H,1,4,6-11H2,2-3H3/b13-5+/t14-,15-,16+,17-,19+,20+/m1/s1

InChI key

BOJKULTULYSRAS-OTESTREVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Andrographolide, a bioactive diterpene lactone, is the main constituent of Andrographis paniculata, a plant used in traditional medicines. It is known to have different biological activities such as anti-inflammatory[1], anti-malarial, anti-cancer[2] and hepatoprotective activity.[3] It also shows potent anti-viral effect against dengue virus.[4]

Application


  • Andrographolide-based drug candidate against Keap1-Nrf2 pathway through rigorous cheminformatics screening.: This study identifies potential andrographolide derivatives as innovative drug candidates that target the Keap1-Nrf2 pathway, crucial for antioxidant defense mechanisms. This approach highlights the compound′s versatility in developing treatments that could mitigate oxidative stress-related diseases, making it a significant area of interest for chemists and material scientists in both academia and biotech sectors (Jain and Sudandira Doss, 2023).

  • TMT-based quantitative proteomics reveals the targets of andrographolide on LPS-induced liver injury.: This investigation into the proteomic changes induced by andrographolide in liver injury models offers insights into its therapeutic mechanisms, highlighting its potential as a treatment for inflammation and liver diseases, relevant for research and development in pharmaceutical and biochemical fields (Ge et al., 2023).

  • Investigation of targets and anticancer mechanisms of covalently acting natural products by functional proteomics.: This study evaluates andrographolide′s roles within the context of its anticancer properties, providing a detailed analysis of its interactions at the molecular level, essential for material scientists and chemists working on novel cancer therapies (Zhao et al., 2023).

Biochem/physiol Actions

Diterpenoid lactone with anti-inflammatory properties. It blocks T-cell proliferation to allogenic stimuli and the chemotactic migration of macrophages induced by complement. Blocks the proliferation of several cancer cell lines in vitro.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

In vitro and in vivo anti-inflammatory effects of andrographolide.
Abu-Ghefreh Ala'AA, et al.
International Immunopharmacology, 9(3), 313-318 (2009)
Hepatoprotective activity of andrographolide from Andrographis paniculata against carbontetrachloride.
Handa SS and Sharma A.
The Indian Journal of Medical Research, 92, 276-283 (1990)
Activity of andrographolide against dengue virus.
Panraksa P, et al.
Antiviral Research, 139, 69-78 (2017)
Andrographolide, a potential cancer therapeutic agent isolated from Andrographis paniculata.
Rajagopal S, et al.
Journal of Experimental Therapeutics & Oncology, 3(3), 147-158 (2003)
Jian Gao et al.
Cell death & disease, 10(12), 957-957 (2019-12-22)
Radiation-induced lung injury (RILI) is one of the most common and fatal complications of thoracic radiotherapy, whereas no effective interventions are available. Andrographolide, an active component extracted from Andrographis paniculate, is prescribed as a treatment for upper respiratory tract infection.

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is the andrographolide can be detected by spectrophotometry UV-VIS ???

    1 answer
    1. Due to the presence of pi bonds in the molecular structure, Andrographolide has chromophores. As a result, this compound may be detected by UV-Vis spectrophotometry.
      Please see the link below to review a publication that may be helpful:
      https://www.pharmahealthsciences.net/pdfs/volume7-issue12019/8.vol7-issue1-2019-MS-15711.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service