Skip to Content
Merck
All Photos(1)

Documents

54750

Sigma-Aldrich

Methyl 4-hydroxybenzoate

puriss., ≥99.0% (GC)

Synonym(s):

p-Hydroxybenzoic acid methyl ester, Methyl paraben, NIPAGIN

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
509801
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

puriss.

Assay

≥99.0% (GC)

ign. residue

≤0.1% (as SO4)

mp

125-128 °C (lit.)
125-128 °C

SMILES string

COC(=O)c1ccc(O)cc1

InChI

1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3

InChI key

LXCFILQKKLGQFO-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

334.4 °F

Flash Point(C)

168 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyung-Jin Min et al.
Aging cell, 7(2), 199-206 (2008-01-29)
Reduced insulin/insulin-like growth factor (IGF) signaling may be a natural way for the reduction of dietary nutrients to extend lifespan. While evidence challenging this hypothesis is accumulating with Caenorhabditis elegans, for Drosophila melanogaster it is still thought that insulin/IGF and
San-Yong Wang et al.
Bioorganic & medicinal chemistry letters, 17(10), 2785-2788 (2007-03-28)
Twelve allenic aromatic ethers, some of them are natural products isolated from the mangrove fungus Xylaria sp. 2508 in the South China Sea, were synthesized. Their antitumor activities against KB and KBv200 cells were determined. All these compounds demonstrated cytotoxic
Peng-Cheng Lv et al.
European journal of medicinal chemistry, 44(4), 1779-1787 (2008-06-10)
Twenty-six depsides were synthesized to screen for their antibacterial activity. All of them were reported for the first time. Their chemical structures were clearly determined by (1)H NMR, (13)C NMR, ESI mass spectra and elemental analyses, coupled with one selected
Fabiana Manservisi et al.
Reproductive toxicology (Elmsford, N.Y.), 54, 110-119 (2015-01-03)
The aim of the study is to determine whether low doses of "endocrine disrupting chemicals" (EDCs) affect the development and proliferative activity of the mammary glands (MGs). Adult parous/nulliparous female Sprague-Dawley (SD) rats were treated from post-natal day (PND) 1
Peter J McNamara et al.
Antimicrobial agents and chemotherapy, 53(5), 1898-1906 (2009-02-19)
Menstrual toxic shock syndrome is a rare but potentially life-threatening illness manifest through the actions of Staphylococcus aureus toxic shock syndrome toxin 1 (TSST-1). Previous studies have shown that tampon additives can influence staphylococcal TSST-1 production. We report here on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service