Merck
  • Effect of maternal exposure to endocrine disrupting chemicals on reproduction and mammary gland development in female Sprague-Dawley rats.
Effect of maternal exposure to endocrine disrupting chemicals on reproduction and mammary gland development in female Sprague-Dawley rats.
Reproductive toxicology (Elmsford, N.Y.) (2015-01-03)
Fabiana Manservisi, Kalpana Gopalakrishnan, Eva Tibaldi, Albana Hysi, Manuela Iezzi, Luca Lambertini, Susan Teitelbaum, Jia Chen, Fiorella Belpoggi
ABSTRACT
The aim of the study is to determine whether low doses of "endocrine disrupting chemicals" (EDCs) affect the development and proliferative activity of the mammary glands (MGs). Adult parous/nulliparous female Sprague-Dawley (SD) rats were treated from post-natal day (PND) 1 until PND 180 with diethylphthalate (DEP), methylparaben (MPB), triclosan (TCS) and a mixture at doses comparable to human exposure. The doses (mg/kg b.w./day) were: DEP=0.173; MPB=0.105; TCS=0.05. EDC treatment resulted in mortality rates >20% in pups as early as lactation day 7. Significant morphological/histological changes were observed at the end of lactation in the MGs of EDC-treated dams. The total transcriptome profile as well as lactation-related genes in MGs also corroborate the morphological findings as more profound gene expression changes are present only at the weaning period. The study highlights the heightened sensitivity of the MGs during critical windows of exposure, particularly pregnancy and lactation, with an impact on pups' survival.
Sigma-Aldrich
Methyl salicylate, ≥98%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methyl salicylate, ReagentPlus®, ≥99% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ≥99%, FCC
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Methyl 4-hydroxybenzoate, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
Methyl 4-hydroxybenzoate, analytical standard
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis